MAT390503386 Matrix Scientific

Benzylidenebis(tricyclohexylphosphine)-dichlororuthenium

  • MDL number MFCD01074452
  • Cas Number : 172222-30-9
  • Molecular Weight : 822.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/092054.html

Safety Information

MSDS http://www.matrixscientific.com/092054.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart