MAT390503697 Matrix Scientific

(6R,7R)-Benzhydryl 3-hydroxy-8-oxo-7-(2-phenylacetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

  • MDL number MFCD00191259
  • Cas Number : 54639-48-4
  • Molecular Weight : 500.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/092365.html

Safety Information

MSDS http://www.matrixscientific.com/092365.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart