MAT390503844 Matrix Scientific

4-Nitrobenzene-1,2-diamine sulfate

  • MDL number MFCD00136899
  • Cas Number : 68239-82-7
  • Molecular Weight : 251.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/092512.html

Safety Information

MSDS http://www.matrixscientific.com/092512.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart