MAT390504516 Matrix Scientific

(3-(Cyclopropylmethoxy)phenyl)boronic acid

  • MDL number MFCD04039016
  • Cas Number : 411229-76-0
  • Molecular Weight : 192.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/093187.html

Safety Information

MSDS http://www.matrixscientific.com/093187.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart