MAT390504766 Matrix Scientific

(2S,4S)-4-Fluoropyrrolidine-2-carboxylic acid

  • MDL number MFCD02683983
  • Cas Number : 2438-57-5
  • Molecular Weight : 133.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/093437.html

Safety Information

MSDS http://www.matrixscientific.com/093437.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart