MAT390505152 Matrix Scientific

Fmoc-Arg(Boc)2-OH

  • MDL number MFCD00237008
  • Cas Number : 143824-77-5
  • Molecular Weight : 596.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.matrixscientific.com/093824.html

Safety Information

MSDS http://www.matrixscientific.com/093824.html
pictograms Irritant/Store in Freezer
Availability details for
Quantity
  • Availability
  • Add to Cart