MAT390505160 Matrix Scientific

H-Tyr(Bzl)-Ome HCl

  • MDL number MFCD00080888
  • Cas Number : 34805-17-9
  • Molecular Weight : 321.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/093832.html

Safety Information

MSDS http://www.matrixscientific.com/093832.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart