MAT390505197 Matrix Scientific

Naphtho[2,3-c]furan-1,3-dione

  • MDL number MFCD00059091
  • Cas Number : 716-39-2
  • Molecular Weight : 198.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 248
Purity 95
Storage temp. Ambient/argon
Country of Origin USA
Boiling point 380
Product Link http://www.matrixscientific.com/093869.html

Safety Information

MSDS http://www.matrixscientific.com/093869.html
pictograms Irritant/Moisture Sensitive/Stored under Argon
Availability details for
Quantity
  • Availability
  • Add to Cart