MAT390505310 Matrix Scientific

2-Dodecylbenzenesulfonic acid

  • MDL number MFCD00066675
  • Cas Number : 27176-87-0
  • Molecular Weight : 326.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 10
Purity 95
Density 1.0/25
Flash Point 85
Storage temp. Freezer
Country of Origin USA
Boiling point 315
Product Link http://www.matrixscientific.com/093982.html

Safety Information

MSDS http://www.matrixscientific.com/093982.html
pictograms Irritant/Store in Freezer
Availability details for
Quantity
  • Availability
  • Add to Cart