MAT390505328 Matrix Scientific

N-(4-Chlorophenyl)-2,6-difluorobenzamide

  • MDL number MFCD04804687
  • Cas Number : 122987-01-3
  • Molecular Weight : 267.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/094000.html

Safety Information

MSDS http://www.matrixscientific.com/094000.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart