MAT390505531 Matrix Scientific

2-Hydroxy-3,5-dinitro-N'-((5-nitrofuran-2-yl)methylene)benzohydrazide

  • MDL number MFCD00377430
  • Cas Number : 16915-70-1
  • Molecular Weight : 365.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/094203.html

Safety Information

MSDS http://www.matrixscientific.com/094203.html
pictograms Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart