MAT390505536 Matrix Scientific

(R)-4-Nitrophenyl 2-((tert-butoxycarbonyl)-amino)-3-phenylpropanoate

  • MDL number MFCD00069874
  • Cas Number : 16159-70-9
  • Molecular Weight : 386.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/094208.html

Safety Information

MSDS http://www.matrixscientific.com/094208.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart