MAT390505585 Matrix Scientific

Boc-Met-ONp

  • MDL number MFCD00038123
  • Cas Number : 2488-18-8
  • Molecular Weight : 370.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/094257.html

Safety Information

MSDS http://www.matrixscientific.com/094257.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart