MAT390506097 Matrix Scientific

2-Acetamido-2-(4-octylphenethyl)-propane-1,3-diyl diacetate

  • MDL number MFCD09835537
  • Cas Number : 162358-09-0
  • Molecular Weight : 433.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Density 1.043
Flash Point 305
Country of Origin USA
Boiling point 582
Product Link http://www.matrixscientific.com/094779.html

Safety Information

MSDS http://www.matrixscientific.com/094779.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart