MAT390506256 Matrix Scientific

2,2'-(1,4-Phenylene)bis(4H-benzo[d][1,3]oxazin-4-one)

  • MDL number MFCD00356293
  • Cas Number : 18600-59-4
  • Molecular Weight : 368.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/094939.html

Safety Information

MSDS http://www.matrixscientific.com/094939.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart