MAT390506540 Matrix Scientific

3-Iodo-6-nitro-1H-indazole-4-carboxylic acid

  • MDL number MFCD07781681
  • Cas Number : 885519-69-7
  • Molecular Weight : 333.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/095224.html

Safety Information

MSDS http://www.matrixscientific.com/095224.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart