MAT390506546 Matrix Scientific

3-Iodo-4-nitro-1H-indazole-6-carboxylic acid

  • MDL number MFCD07781727
  • Cas Number : 885521-14-2
  • Molecular Weight : 333.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/095230.html

Safety Information

MSDS http://www.matrixscientific.com/095230.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart