MAT390506559 Matrix Scientific

(S)-Di-tert-Butyl 2-(2-hydroxyethyl)-piperazine-1,4-dicarboxylate

  • MDL number MFCD20489327
  • Cas Number : 660862-48-6
  • Molecular Weight : 330.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/095243.html

Safety Information

MSDS http://www.matrixscientific.com/095243.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart