MAT390506697 Matrix Scientific

Trimethylolpropane trioleate

  • MDL number MFCD00072630
  • Cas Number : 57675-44-2
  • Molecular Weight : 827.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/095382.html

Safety Information

MSDS http://www.matrixscientific.com/095382.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart