MAT390507141 Matrix Scientific

9,9-Diphenyl-9H-fluoren-2-amine

  • MDL number MFCD22376664
  • Cas Number : 1268519-74-9
  • Molecular Weight : 333.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/095826.html

Safety Information

msds http://www.matrixscientific.com/095826.html
Availability details for
Quantity
  • Availability
  • Add to Cart