MAT390507312 Matrix Scientific

Iridium(IV) chloride

  • MDL number MFCD00016155
  • Cas Number : 10025-97-5
  • Molecular Weight : 334.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point >350
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/095997.html

Safety Information

MSDS http://www.matrixscientific.com/095997.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart