MAT390507515 Matrix Scientific

3-Fluoropropyl 4-methylbenzenesulfonate

  • MDL number MFCD01658062
  • Cas Number : 312-68-5
  • Molecular Weight : 232.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Boiling point 84�mHg
Product Link http://www.matrixscientific.com/096201.html

Safety Information

MSDS http://www.matrixscientific.com/096201.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart