MAT390507658 Matrix Scientific

4-Chloro-6-fluoropyrido[3,4-d]pyrimidine

  • MDL number MFCD09037872
  • Cas Number : 175357-98-9
  • Molecular Weight : 183.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/096344.html

Safety Information

MSDS http://www.matrixscientific.com/096344.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart