MAT390507943 Matrix Scientific

N-Lauroyl-L-lysine

  • MDL number MFCD00216730
  • Cas Number : 52315-75-0
  • Molecular Weight : 328.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/096629.html

Safety Information

MSDS http://www.matrixscientific.com/096629.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart