MAT390507956 Matrix Scientific

10-(3-Chloropropyl)-2-(trifluoromethyl)-10H-phenothiazine

  • MDL number MFCD08549622
  • Cas Number : 1675-46-3
  • Molecular Weight : 343.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 74-76
Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/096642.html

Safety Information

MSDS http://www.matrixscientific.com/096642.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart