MAT390508008 Matrix Scientific

4,7-Bis(4-bromophenyl)-1,10-phenanthroline

  • MDL number MFCD22493502
  • Cas Number : 97802-08-9
  • Molecular Weight : 490.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/096694.html

Safety Information

MSDS http://www.matrixscientific.com/096694.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart