MAT390508025 Matrix Scientific

(3-((2-(Dimethylamino)ethyl)carbamoyl)-phenyl)boronic acid

  • MDL number MFCD04115706
  • Cas Number : 850567-31-6
  • Molecular Weight : 236.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.matrixscientific.com/096711.html

Safety Information

MSDS http://www.matrixscientific.com/096711.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart