MAT390508108 Matrix Scientific

5,5-Dimethyldihydrothiophen-3(2H)-one

  • MDL number MFCD13195297
  • Cas Number : 32858-41-6
  • Molecular Weight : 130.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/096794.html

Safety Information

MSDS http://www.matrixscientific.com/096794.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart