MAT390508169 Matrix Scientific

2-Chloro-5-fluoronicotinamide

  • MDL number MFCD16658704
  • Cas Number : 75302-64-6
  • Molecular Weight : 174.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/096855.html

Safety Information

MSDS http://www.matrixscientific.com/096855.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart