MAT390508206 Matrix Scientific

4-(((1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl)-oxy)-4-oxobutanoic acid

  • MDL number MFCD23135646
  • Cas Number : 77341-67-4
  • Molecular Weight : 256.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/096892.html

Safety Information

MSDS http://www.matrixscientific.com/096892.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart