MAT390508630 Matrix Scientific

(5-(((tert-Butoxycarbonyl)amino)methyl)-pyridin-3-yl)boronic acid

  • MDL number MFCD07374885
  • Cas Number : 1309981-05-2
  • Molecular Weight : 252.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/097316.html

Safety Information

MSDS http://www.matrixscientific.com/097316.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart