MAT390508720 Matrix Scientific

5'-O-Trityluridine

  • MDL number MFCD00038051
  • Cas Number : 6554-10-5
  • Molecular Weight : 486.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207-208
Purity 98
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/097406.html

Safety Information

MSDS http://www.matrixscientific.com/097406.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart