MAT390508844 Matrix Scientific

Naphthalene-1,4,5,8-tetracarboxylic acid

  • MDL number MFCD00004012
  • Cas Number : 128-97-2
  • Molecular Weight : 304.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/097531.html

Safety Information

MSDS http://www.matrixscientific.com/097531.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart