MAT390508908 Matrix Scientific

2-((Tribromomethyl)sulfonyl)pyridine

  • MDL number MFCD02093493
  • Cas Number : 59626-33-4
  • Molecular Weight : 393.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/097595.html

Safety Information

MSDS http://www.matrixscientific.com/097595.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart