MAT390508909 Matrix Scientific

3-Aminophenol sulfate (2:1)

  • MDL number MFCD08460117
  • Cas Number : 68239-81-6
  • Molecular Weight : 316.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/097596.html

Safety Information

MSDS http://www.matrixscientific.com/097596.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart