MAT390508937 Matrix Scientific

3,3-Bis(bromomethyl)pentane

  • MDL number MFCD00039190
  • Cas Number : 67969-84-0
  • Molecular Weight : 258.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 38-41
Purity 90
Country of Origin USA
Product Link http://www.matrixscientific.com/097624.html

Safety Information

MSDS http://www.matrixscientific.com/097624.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart