MAT390509209 Matrix Scientific

1-(2,6-Difluorobenzyl)-1H-1,2,3-triazole-4-carbonitrile

  • MDL number MFCD09859711
  • Cas Number : 202003-06-3
  • Molecular Weight : 220.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/097896.html

Safety Information

MSDS http://www.matrixscientific.com/097896.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart