MAT390509253 Matrix Scientific

Sodium 3-oxo-3H-benzo[d]isothiazol-2-ide 1,1-dioxide

  • MDL number MFCD00040618
  • Cas Number : 128-44-9
  • Molecular Weight : 205.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/097940.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/097940.html
Availability details for
Quantity
  • Availability
  • Add to Cart