MAT390509257 Matrix Scientific

Sorbitan tristearate

  • MDL number MFCD00151165
  • Cas Number : 26658-19-5
  • Molecular Weight : 859.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/097944.html

Safety Information

MSDS http://www.matrixscientific.com/097944.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart