MAT390509360 Matrix Scientific

Aniline phosphinate

  • MDL number MFCD07369777
  • Cas Number : 82395-88-8
  • Molecular Weight : 159.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-115
Purity 95
Storage temp. Ambient/argon
Country of Origin USA
Product Link http://www.matrixscientific.com/098048.html

Safety Information

MSDS http://www.matrixscientific.com/098048.html
pictograms Toxic/Stored under Argon
Availability details for
Quantity
  • Availability
  • Add to Cart