MAT390509398 Matrix Scientific

Quinine sulfate

  • MDL number MFCD00135950
  • Cas Number : 804-63-7
  • Molecular Weight : 422.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/098086.html

Safety Information

MSDS http://www.matrixscientific.com/098086.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart