MAT390509429 Matrix Scientific

(S)-2-(4-((((S)-2-Amino-5-methyl-4-oxo-3,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid

  • MDL number MFCD21363683
  • Cas Number : 31690-09-2
  • Molecular Weight : 459.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/098117.html

Safety Information

MSDS http://www.matrixscientific.com/098117.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart