MAT390509472 Matrix Scientific

(1a)-17-(Acetyloxy)-6-chloro-1-(chloromethyl)-pregna-4,6-diene-3,20-dione

  • MDL number MFCD09833378
  • Cas Number : 17183-98-1
  • Molecular Weight : 453.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/098160.html

Safety Information

MSDS http://www.matrixscientific.com/098160.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart