MAT390509574 Matrix Scientific

2-(Methylamino)acetonitrile

  • MDL number MFCD00058964
  • Cas Number : 5616-32-0
  • Molecular Weight : 70.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Density 0.92
Storage temp. Freezer
Country of Origin USA
Boiling point 65
Product Link http://www.matrixscientific.com/098262.html

Safety Information

MSDS http://www.matrixscientific.com/098262.html
pictograms Flammable/Toxic/Corrosive/Store in Freezer
Availability details for
Quantity
  • Availability
  • Add to Cart