MAT390509747 Matrix Scientific

N,N,N-Trimethyl-1-phenylmethanaminium iodide

  • MDL number MFCD00051859
  • Cas Number : 4525-46-6
  • Molecular Weight : 277.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-179
Purity 95
Storage temp. Refrigerate/argon/dark
Country of Origin USA
Product Link http://www.matrixscientific.com/098435.html

Safety Information

MSDS http://www.matrixscientific.com/098435.html
pictograms Irritant/Light Sensitive/Hygroscopic/Refrigerate/Stored under Argon
Availability details for
Quantity
  • Availability
  • Add to Cart