MAT390509795 Matrix Scientific

N,N-Dimethyl-N-octyloctan-1-aminium chloride

  • MDL number MFCD01711139
  • Cas Number : 5538-94-3
  • Molecular Weight : 305.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 80
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/098483.html

Safety Information

MSDS http://www.matrixscientific.com/098483.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart