MAT390509812 Matrix Scientific

Antioxidant 1024

  • MDL number MFCD00072662
  • Cas Number : 32687-78-8
  • Molecular Weight : 552.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227-232
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/098500.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/098500.html
Availability details for
Quantity
  • Availability
  • Add to Cart