MAT390510132 Matrix Scientific

Sodium 2-(dimethylamino)acetate

  • MDL number MFCD00082922
  • Cas Number : 18319-88-5
  • Molecular Weight : 125.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/098823.html

Safety Information

MSDS http://www.matrixscientific.com/098823.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart