MAT390510150 Matrix Scientific

Potassium 3-(phenylsulfonyl)benzenesulfonate

  • MDL number MFCD00036079
  • Cas Number : 63316-43-8
  • Molecular Weight : 336.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/098841.html

Safety Information

MSDS http://www.matrixscientific.com/098841.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart