MAT390510158 Matrix Scientific

Pyranthrene-8,16-dione

  • MDL number MFCD00049379
  • Cas Number : 128-70-1
  • Molecular Weight : 406.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/098849.html

Safety Information

MSDS http://www.matrixscientific.com/098849.html
pictograms Irritant
Availability details for
Quantity
  • Availability
  • Add to Cart